Tetrafluoroboric acid ≥48% (w/w) in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Fluoboric acid
11484-18
11484-36
11484-57
CAAA11484-18EA
87.22
CAD
CAAA11484-18
CAAA11484-36
CAAA11484-57
Tetrafluoroboric acid ≥48% (w/w) in aqueous solution
Tetrafluoroboric acid
MDL: MFCD00011345
CAS No.: 16872-11-0
Molecular Formula: HBF4
Formula Weight: 87.81
Physical Form: Liquid
Density: 1.4
Appearance: Colorless to pale yellow
Odor: Pungent
Boiling Point: 130° dec
Merck Reference: 14,4149
UN No.: 1775
MDL No.: MFCD00011345
Formula:
HBF₄ MW: 87.81 g/mol Boiling Pt: 130 °C (dec.) Density: 1.4 Storage Temperature: Ambient |
MDL Number:
MFCD00011345 CAS Number: 16872-11-0 EINECS: 240-898-3 UN: 1775 ADR: 8,II Merck Index: 14,4149 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...