Vanadium ≥99.8% (metals basis), wire, Ø 0.127 mm (0.005 in)
Supplier: Thermo Scientific Chemicals
11483-G1
11483-G5
CAAA11483-G1EA
124.03
CAD
CAAA11483-G1
CAAA11483-G5
Vanadium ≥99.8% (metals basis), wire, Ø 0.127 mm (0.005 in)
Vanadium
MDL: MFCD00011453
Purity: 99.8%
CAS No.: 7440-62-2
Physical Form: ≈0.08g/m
Odor: Odorless
MDL No.: MFCD00011453
Formula:
V MW: 50.94 g/mol Boiling Pt: 3477 Melting Pt: 1896 Density: 6.015 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011453 CAS Number: 7440-62-2 EINECS: 231-171-1 Merck Index: 12,10053 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...