Aluminium ≥99.9965% (metals basis), rods, Puratronic®, Ø 6 mm (0.2 in)
Supplier: Thermo Scientific Chemicals
11478-BT
11478-QY
CAAA11478-BTEA
214.88
CAD
CAAA11478-BT
CAAA11478-QY
Aluminium ≥99.9965% (metals basis), rods, Puratronic®, Ø 6 mm (0.2 in)
Aluminium
MDL: MFCD00134029
Purity: 99.9965%
CAS No.: 7429-90-5
Physical Form: ≈0.76g/cm
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al Boiling Pt: 2327 °C (1013 hPa) Melting Pt: 660 °C Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...