Molybdenum ≥99.97% (metals basis), rods, Puratronic®, Ø 5 mm (0.2 in)
Supplier: Thermo Scientific Chemicals
11475-DO
11475-FN
CAAA11475-DOEA
251.22
CAD
CAAA11475-DO
CAAA11475-FN
Molybdenum ≥99.97% (metals basis), rods, Puratronic®, Ø 5 mm (0.2 in)
Molybdenum
MDL: MFCD00003465
Purity: 99.97%
CAS No.: 7439-98-7
Physical Form: ≈2g/cm
MDL No.: MFCD00003465
Formula:
Mo Boiling Pt: 3617 °C (1013 hPa) Melting Pt: 2622 °C Density: 10.28 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003465 CAS Number: 7439-98-7 EINECS: 231-107-2 Merck Index: 12,06317 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...