Tin ≥99.9985% (metals basis), rods, Puratronic®, Length: 160 mm, Ø 6 mm (0.2 in)
Supplier: Thermo Scientific Chemicals
11472-DP
11472-HS
CAAA11472-DPEA
197.5
CAD
CAAA11472-DP
CAAA11472-HS
Tin ≥99.9985% (metals basis), rods, Puratronic®, Length: 160 mm, Ø 6 mm (0.2 in)
Tin
MDL: MFCD00133862
Purity: 99.9985%
CAS No.: 7440-31-5
Physical Form: ≈2.06g/cm
Odor: Odorless
MDL No.: MFCD00133862
Formula:
Sn MW: 118.71 g/mol Boiling Pt: 2270 Melting Pt: 232 Density: 7.31 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 Merck Index: 12,09587 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...