Silver 99,9985 Metals basis, wire, Premion®, Ø 0.5 mm (0.02 in)
Supplier: Thermo Scientific Chemicals
11469-G1
11469-G5
11469-G9
CAAA11469-G1EA
91.96
CAD
CAAA11469-G1
CAAA11469-G5
CAAA11469-G9
Silver 99,9985 Metals basis, wire, Premion®, Ø 0.5 mm (0.02 in)
Silver
MDL: MFCD00003397
Purity: 99.9985%
CAS No.: 7440-22-4
Physical Form: ≈2.06g/m
Odor: Characteristic
MDL No.: MFCD00003397
Formula:
Ag MW: 107.87 g/mol Boiling Pt: 2212 Melting Pt: 962 Density: 10.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003397 CAS Number: 7440-22-4 EINECS: 231-131-3 Merck Index: 12,08647 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...