Silver 99,999 Metals basis, wire, Premion®, Ø 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

11466-BW 11466-CB 11466-CD
CAAA11466-BWEA 140.15 CAD
CAAA11466-BW CAAA11466-CB CAAA11466-CD
Silver 99,999 Metals basis, wire, Premion®, Ø 1.0 mm (0.04 in)
Silver

MDL: MFCD00003397


Purity: 99.999% CAS No.: 7440-22-4 Physical Form: ≈8.24g/m Odor: Characteristic MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR