Tin ≥99.998% (metals basis), foil, Puratronic®, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
11459-FI
11459-GH
11459-HP
CAAA11459-FIEA
112.18
CAD
CAAA11459-FI
CAAA11459-GH
CAAA11459-HP
Tin ≥99.998% (metals basis), foil, Puratronic®, Thickness: 0.25 mm (0.01 in)
Tin
MDL: MFCD00133862
Purity: 99.998%
CAS No.: 7440-31-5
Physical Form: ≈4.6g/50x50mm
Odor: Odorless
MDL No.: MFCD00133862
Formula:
Sn Boiling Pt: 2270 °C (1013 hPa) Melting Pt: 232 °C Density: 7.31 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 Merck Index: 12,09587 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...