Tin ≥99.9985% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Supplier: Thermo Scientific Chemicals
11458-FI
11458-GH
CAAA11458-FIEA
162.74
CAD
CAAA11458-FI
CAAA11458-GH
Tin ≥99.9985% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Tin
MDL: MFCD00133862
Purity: 99.9985%
CAS No.: 7440-31-5
Physical Form: ≈9.1g/50x50mm
Odor: Odorless
MDL No.: MFCD00133862
Formula:
Sn Boiling Pt: 2270 °C (1013 hPa) Melting Pt: 232 °C Density: 7.31 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 Merck Index: 12,09587 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...