Tin ≥99.9985% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

11457-FG 11457-FY 11457-HL
CAAA11457-FGEA 122.92 CAD
CAAA11457-FG CAAA11457-FY CAAA11457-HL
Tin ≥99.9985% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)
Tin

MDL: MFCD00133862


Purity: 99.9985% CAS No.: 7440-31-5 Physical Form: ≈9.1g/25x50mm Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR