Iron ≥99.995% (metals basis), rods, Puratronic®, Ø 5 mm (0.2 in)

Supplier: Thermo Scientific Chemicals

11442-DJ 11442-DO 11442-HT
CAAA11442-DJEA 366.56 CAD
CAAA11442-DJ CAAA11442-DO CAAA11442-HT
Iron ≥99.995% (metals basis), rods, Puratronic®, Ø 5 mm (0.2 in)
Iron

MDL: MFCD00010999


Purity: 99.995% CAS No.: 7439-89-6 Physical Form: ≈0.155g/mm MDL No.: MFCD00010999

Formula: Fe
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1535 °C
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR