Silver ≥99.9% (metals basis), shot 1-5 mm (0.04-0.2 in)

Supplier: Thermo Scientific Chemicals

11436-09 11436-18 11436-30
CAAA11436-09EA 122.45 CAD
CAAA11436-09 CAAA11436-18 CAAA11436-30
Silver ≥99.9% (metals basis), shot 1-5 mm (0.04-0.2 in)
Silver

MDL: MFCD00003397


Purity: 99.9% CAS No.: 7440-22-4 Odor: Characteristic MDL No.: MFCD00003397

Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR