Silver hard_SC 99,9 Metals basis, wire 1/2 hard, hard, Ø 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

11433-G2 11433-G6 11433-H2
CAAA11433-G2EA 79.63 CAD
CAAA11433-G2 CAAA11433-G6 CAAA11433-H2
Silver hard_SC 99,9 Metals basis, wire 1/2 hard, hard, Ø 0.5 mm (0.02 in)
Silver

MDL: MFCD00003397


Purity: 99.9% CAS No.: 7440-22-4 Physical Form: ≈2.06g/m, Temper: 1/2 hard Odor: Characteristic MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR