Silver cyanide 99%

Supplier: Thermo Scientific Chemicals
Danger

11424-14 11424-22
CAAA11424-14EA 251.22 CAD
CAAA11424-14 CAAA11424-22
Silver cyanide 99%
Silver cyanide

Silver plating


Purity: 99% CAS No.: 506-64-9 Molecular Formula: CAgN Formula Weight: 133.84 Physical Form: Powder Density: 3.95 Refractive Index: 1.685 Sensitivity: Light Sensitive Appearance: White to off-white Odor: Odorless Melting Point: 320° dec. Solubility: Insoluble in water, dilute acids, and alcohol. Soluble in alkali cyanides and boiling concentrated HNO3 Merck Reference: 14,8512 UN No.: 1684 MDL No.: MFCD00003409


Silver plating

Formula: AgCN
Melting Pt: 320 °C (dec.)
Density: 3.95
Storage Temperature: Ambient
MDL Number: MFCD00003409
CAS Number: 506-64-9
EINECS: 208-048-6
UN: 1684
ADR: 6.1,II
Merck Index: 14,8512

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR