Silver chloride ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
11421-14
11421-22
11421-36
CAAA11421-14EA
270.18
CAD
CAAA11421-14
CAAA11421-22
CAAA11421-36
Silver chloride ≥99.9% (metals basis)
Silver chloride
MDL: MFCD00003399
Purity: 99.9%
CAS No.: 7783-90-6
Molecular Formula: AgCl
Formula Weight: 143.32
Physical Form: Crystalline
Density: 5.56
Refractive Index: 2.071
Sensitivity: Light Sensitive
Appearance: White
Odor: Odorless
Melting Point: 455°
Boiling Point: 1550°
Merck Reference: 14,8509
UN No.: 3077
MDL No.: MFCD00003399
Formula:
AgCl MW: 143.32 g/mol Boiling Pt: 1554 Melting Pt: 455 Density: 5.56 Storage Temperature: Ambient |
MDL Number:
MFCD00003399 CAS Number: 7783-90-6 EINECS: 232-033-3 UN: 3077 ADR: 9 III Merck Index: 14,8509 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...