Silver carbonate ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
11420-14
11420-22
CAAA11420-14EA
208.56
CAD
CAAA11420-14
CAAA11420-22
Silver carbonate ≥99.5% (metals basis)
Silver carbonate
MDL: MFCD00003403
Insoluble in water. Freely soluble in dilute HNO3, ammonia
Purity: 99.5%
CAS No.: 534-16-7
Molecular Formula: CAg2O3
Formula Weight: 275.75
Physical Form: Powder
Density: 6.08
Appearance: Yellow
Odor: Odorless
Melting Point: 218° dec.
Solubility: Insoluble in water. Freely soluble in dilute HNO3, ammonia
Merck Reference: 14,8507
UN No.: 3077
MDL No.: MFCD00003403
Formula:
CAg₂O₃ MW: 275.75 g/mol Melting Pt: 218 °C (dec.) Density: 6.08 Storage Temperature: Ambient |
MDL Number:
MFCD00003403 CAS Number: 534-16-7 EINECS: 208-590-3 UN: 3077 ADR: 9 III Merck Index: 14,8507 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...