Silver iodide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

11419-14 11419-22
CAAA11419-14EA 265.44 CAD
CAAA11419-14 CAAA11419-22
Silver iodide ≥99.9% (metals basis)
Silver iodide

MDL: MFCD00003412 Insoluble in water, acids, ammonium carbonate. Freely soluble in alkali cyanides and iodides.


Purity: 99.9% CAS No.: 7783-96-2 Molecular Formula: AgI Formula Weight: 234.77 Physical Form: Crystalline Density: 5.67 Appearance: Yellow Odor: Odorless Melting Point: 558° Solubility: Insoluble in water, acids, ammonium carbonate. Freely soluble in alkali cyanides and iodides Merck Reference: 14,8516 UN No.: 3077 MDL No.: MFCD00003412

Formula: AgI
Boiling Pt: 1506 °C (1013 hPa)
Melting Pt: 558 °C
Density: 5.67
Storage Temperature: Ambient
MDL Number: MFCD00003412
CAS Number: 7783-96-2
EINECS: 232-038-0
UN: 3077
ADR: 9,III
Merck Index: 14,8516

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR