Silver nitrite ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
11418-09
11418-18
CAAA11418-09EA
170.64
CAD
CAAA11418-09
CAAA11418-18
Silver nitrite ≥99% (metals basis)
Silver nitrite
MDL: MFCD00003413
Purity: 99%
CAS No.: 7783-99-5
Molecular Formula: AgNO2
Formula Weight: 153.88
Physical Form: Crystalline
Density: 4.45
Appearance: Pale yellow
Odor: Odorless
Melting Point: 140° dec.
Merck Reference: 14,8519
UN No.: 2627
MDL No.: MFCD00003413
Formula:
AgNO₂ Melting Pt: 140 °C (dec.) Density: 4.45 Storage Temperature: Ambient |
MDL Number:
MFCD00003413 CAS Number: 7783-99-5 EINECS: 232-041-7 UN: 2627 ADR: 5.1,II Merck Index: 14,8519 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...