Silver sulfate ≥98% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Disilver sulfate, Silver(I) sulfate, Disilver sulphate

11417-14 11417-22
CAAA11417-14EA 199.08 CAD
CAAA11417-14 CAAA11417-22
Silver sulfate ≥98% ACS
Silver sulphate

MDL: MFCD00003407 Partially soluble in water. Water solubility increases with temperature. Soluble in HNO3, ammonia, conc. H2SO3


Purity: 98% CAS No.: 10294-26-5 Molecular Formula: Ag2O4S Formula Weight: 311.80 Physical Form: Powder Density: 5.45 Appearance: White Odor: Odorless Melting Point: 657° Boiling Point: 1085° dec. Solubility: Partially soluble in water. Water solubility increases with temperature. Soluble in HNO3, ammonia, conc. H2SO4 Merck Reference: 14,8529 UN No.: 3077 MDL No.: MFCD00003407

Formula: Ag₂SO₄
MW: 311.8 g/mol
Boiling Pt: 1085 (dec.)
Melting Pt: 655
Density: 5.45
Storage Temperature: Ambient
MDL Number: MFCD00003407
CAS Number: 10294-26-5
EINECS: 233-653-7
Merck Index: 14,8529
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Insoluble matter and silver chloride 0.02%
NO3 0.1%
Substances not precipitated by hydrochloric acid 0.03%
Fe 0.00001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR