Perrhenic acid ≥75-80% in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

11413-06 11413-14
CAAA11413-06EA 206.98 CAD
CAAA11413-06 CAAA11413-14
Perrhenic acid ≥75-80% in aqueous solution
Perrhenic acid

MDL: MFCD00011326


CAS No.: 13768-11-1 Molecular Formula: HO4Re Formula Weight: 251.26 Density: 2.42 Appearance: Colorless to pale yellow UN No.: 3264 MDL No.: MFCD00011326

Formula: HO₄Re
Density: 2.42
Storage Temperature: Ambient
MDL Number: MFCD00011326
CAS Number: 13768-11-1
EINECS: 237-380-4
UN: 3264
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR