Potassium perrhenate (64% Re) ≥99% (metals basis) Re 64%

Supplier: Thermo Scientific Chemicals
Danger

11411-04 11411-09 11411-18
CAAA11411-04EA 107.44 CAD
CAAA11411-04 CAAA11411-09 CAAA11411-18 CAAA11411-SS
Potassium perrhenate (64% Re) ≥99% (metals basis) Re 64%
Potassium perrhenate

A powerful oxidizer


Purity: 99% CAS No.: 10466-65-6 Molecular Formula: KReO4 Formula Weight: 289.30 Physical Form: Crystalline Density: 4.887 Refractive Index: 1.643 Appearance: White Odor: Odorless Melting Point: 550° Boiling Point: 1360-1370° UN No.: 1479 MDL No.: MFCD00011365


A powerful oxidizer

Formula: KO₄Re
Boiling Pt: 1360…1370 °C
Melting Pt: 550 °C
Density: 4,887
Storage Temperature: Ambient
MDL Number: MFCD00011365
CAS Number: 10466-65-6
EINECS: 233-953-8
UN: 1479
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR