Ammonium perrhenate ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
11410-04
11410-09
11410-18
CAAA11410-04EA
83.58
CAD
CAAA11410-04
CAAA11410-09
CAAA11410-18
Ammonium perrhenate ≥99% (metals basis)
Ammonium perrhenate
MDL: MFCD00012377
Purity: 99+%
CAS No.: 13598-65-7
Molecular Formula: H4NO4Re
Formula Weight: 268.24
Physical Form: Powder
Density: 3.97
Appearance: White
Melting Point: dec.
UN No.: 1479
MDL No.: MFCD00012377
Formula:
H₄NO₄Re Melting Pt: Dec. Density: 3,97 Storage Temperature: Ambient |
MDL Number:
MFCD00012377 CAS Number: 13598-65-7 EINECS: 237-075-6 UN: 1479 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...