Silver ≥99.9% (metals basis), powder APS 4-7 µm

Supplier: Thermo Scientific Chemicals

11402-09 11402-18 11402-30
CAAA11402-09EA 118.66 CAD
CAAA11402-09 CAAA11402-18 CAAA11402-30
Silver ≥99.9% (metals basis), powder APS 4-7 µm
Silver

MDL: MFCD00003397


Purity: 99.9% CAS No.: 7440-22-4 Physical Form: S.A. 0.1-0.4m2/g Odor: Characteristic UN No.: 3077 MDL No.: MFCD00003397

Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 961.9 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR