Strontium titanate 99+% (metals basis)
Supplier: Thermo Scientific Chemicals
11399-30
11399-A1
CAAA11399-30EA
99.22
CAD
CAAA11399-30
CAAA11399-A1
Strontium titanate 99+% (metals basis)
Strontium titanate
Used in electronics and in electrical insulation
Purity: 99+%
CAS No.: 12060-59-2
Molecular Formula: SrTiO3
Formula Weight: 183.51
Physical Form: Powder
Density: 4.810
Melting Point: 2060°
Solubility: Insoluble in water and most solvents
MDL No.: MFCD00049554
Used in electronics and in electrical insulation
Formula:
O₃SrTi Melting Pt: 2060 °C Density: 4,810 Storage Temperature: Ambient |
MDL Number:
MFCD00049554 CAS Number: 12060-59-2 EINECS: 235-044-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...