Magnesium titanate ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals

11398-30 11398-A1
CAAA11398-30EA 69.2 CAD
CAAA11398-30 CAAA11398-A1
Magnesium titanate ≥99% (metals basis)
Magnesium titanate

MDL: MFCD00016198


Purity: 99% CAS No.: 12032-30-3 Molecular Formula: MgTiO3 Formula Weight: 120.21 Physical Form: 2 Micron Powder Density: 3.36 Appearance: White Odor: Odorless Melting Point: 1610° MDL No.: MFCD00016198

Formula: MgO₃Ti
MW: 120.18
Melting Pt: 1610 °C
Density: 3.36
Storage Temperature: Ambient
MDL Number: MFCD00016198
CAS Number: 12032-30-3
EINECS: 234-766-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR