Calcium titanate 99+% (metals basis)
Supplier: Thermo Scientific Chemicals
11397-36
11397-A3
CAAA11397-36EA
200.66
CAD
CAAA11397-36
CAAA11397-A3
Calcium titanate 99+% (metals basis)
Calcium titanate
MDL: MFCD00015981
Purity: 99+%
CAS No.: 12049-50-2
Molecular Formula: CaTiO3
Formula Weight: 135.98
Physical Form: -325 Mesh Powder
Density: 4.10
Melting Point: 1975°
MDL No.: MFCD00015981
Formula:
CaO₃Ti Melting Pt: 1975 °C Density: 4,10 Storage Temperature: Ambient |
MDL Number:
MFCD00015981 CAS Number: 12049-50-2 EINECS: 234-988-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...