Copper ≥99.9985% (metals basis), foil, Puratronic®, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
11393-FI
11393-GH
CAAA11393-FIEA
140.15
CAD
CAAA11393-FI
CAAA11393-GH
Copper ≥99.9985% (metals basis), foil, Puratronic®, Thickness: 0.25 mm (0.01 in)
Copper
MDL: MFCD00010965
Purity: 99.9985%
CAS No.: 7440-50-8
Physical Form: ≈5.6g/50x50mm
Odor: Odorless
MDL No.: MFCD00010965
Formula:
Cu MW: 63.55 g/mol Boiling Pt: 2595 Melting Pt: 1083 Density: 8.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...