Copper ≥99.9985% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

11392-FI 11392-GH
CAAA11392-FIEA 203.82 CAD
CAAA11392-FI CAAA11392-GH
Copper ≥99.9985% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Copper

MDL: MFCD00010965


Purity: 99.9985% CAS No.: 7440-50-8 Physical Form: ≈11.2g/50x50mm Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR