Indium ≥99.9975% (metals basis), foil, Puratronic®, Thickness: 0.1 mm (0.004 in)
Supplier: Thermo Scientific Chemicals
11386-FI
11386-GH
CAAA11386-FIEA
334.96
CAD
CAAA11386-FI
CAAA11386-GH
Indium ≥99.9975% (metals basis), foil, Puratronic®, Thickness: 0.1 mm (0.004 in)
Indium
MDL: MFCD00134048
Purity: 99.9975%
CAS No.: 7440-74-6
Physical Form: ≈1.8g/50x50mm
Odor: Odorless
MDL No.: MFCD00134048
Formula:
In Boiling Pt: 2000 °C (1013 hPa) Melting Pt: 156 °C Density: 7.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...