Indium ≥99.9975% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

11384-FF 11384-FI 11384-FY 11384-GK
CAAA11384-FFEA 130.67 CAD
CAAA11384-FF CAAA11384-FI CAAA11384-FY CAAA11384-GK
Indium ≥99.9975% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Indium

MDL: MFCD00134048


Purity: 99.9975% CAS No.: 7440-74-6 Physical Form: ≈2.3g/25x25mm Odor: Odorless MDL No.: MFCD00134048

Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 156 °C
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR