Indium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 2.0 mm (0.08 in)
Supplier: Thermo Scientific Chemicals
11382-FF
11382-FI
CAAA11382-FFEA
360.24
CAD
CAAA11382-FF
CAAA11382-FI
Indium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 2.0 mm (0.08 in)
Indium
MDL: MFCD00134048
Purity: 99.998%
CAS No.: 7440-74-6
Physical Form: ≈9.1g/25x25mm
Odor: Odorless
MDL No.: MFCD00134048
Formula:
In Boiling Pt: 2000 °C (1013 hPa) Melting Pt: 156 °C Density: 7.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...