Iron ≥99.994% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Supplier: Thermo Scientific Chemicals
11379-FF
11379-FI
11379-GH
CAAA11379-FFEA
197.5
CAD
CAAA11379-FF
CAAA11379-FI
CAAA11379-GH
Iron ≥99.994% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Iron
MDL: MFCD00010999
Purity: 99.994%
CAS No.: 7439-89-6
Physical Form: ≈2.5g/25x25mm
MDL No.: MFCD00010999
Formula:
Fe Boiling Pt: 3000 °C (1013 hPa) Melting Pt: 1535 °C Density: 7.86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010999 CAS Number: 7439-89-6 EINECS: 231-096-4 Merck Index: 13,05109 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...