Aluminium 99,997 Metals basis, foil, Puratronic®, Thickness: 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminum

11376-GH 11376-GL 11376-IP
CAAA11376-GHEA 143.15 CAD
CAAA11376-GH CAAA11376-GL CAAA11376-IP
Aluminium 99,997 Metals basis, foil, Puratronic®, Thickness: 0.1 mm (0.004 in)
Aluminium

MDL: MFCD00134029


Purity: 99.997% CAS No.: 7429-90-5 Physical Form: ≈2.7g/100x100mm, Temper: as rolled Odor: Odorless MDL No.: MFCD00134029

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR