Aluminium ≥99.997% (metals basis), foil, Puratronic®, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
Synonyms:
Aluminum
11375-FI
11375-GH
11375-HP
CAAA11375-FIEA
76.63
CAD
CAAA11375-FI
CAAA11375-GH
CAAA11375-HP
Aluminium ≥99.997% (metals basis), foil, Puratronic®, Thickness: 0.25 mm (0.01 in)
Aluminium
MDL: MFCD00134029
Purity: 99.997%
CAS No.: 7429-90-5
Physical Form: ≈1.7g/50x50mm, Temper: as rolled
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al Boiling Pt: 2327 °C (1013 hPa) Melting Pt: 660 °C Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...