Aluminium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminum

11373-FI 11373-GH 11373-FI 11373-HP 11373-HP 11373-GH
CAAA11373-HPEA 872.16 CAD
AA11373-HP CAAA11373-HP AA11373-FI CAAA11373-FI AA11373-GH CAAA11373-GH
Aluminium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)
Aluminium
Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
Merck Index: 13,00321

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR