Aluminium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 2.0 mm (0.08 in)
Supplier: Thermo Scientific Chemicals
11372-FI
11372-GH
11372-HP
CAAA11372-FIEA
169.06
CAD
CAAA11372-FI
CAAA11372-GH
CAAA11372-HP
Aluminium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 2.0 mm (0.08 in)
Aluminium
MDL: MFCD00134029
Purity: 99.998%
CAS No.: 7429-90-5
Physical Form: ≈13.5g/50x50mm, Temper: as rolled
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al Boiling Pt: 2327 °C (1013 hPa) Melting Pt: 660 °C Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...