Cadmium ≥99.9975% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals
Danger

11369-FI 11369-GH
CAAA11369-FIEA 181.7 CAD
CAAA11369-FI CAAA11369-GH
Cadmium ≥99.9975% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Cadmium

MDL: MFCD00010914


Purity: 99.9975% CAS No.: 7440-43-9 Physical Form: ≈10.8g/50x50mm MDL No.: MFCD00010914

Formula: Cd
Boiling Pt: 765 °C (1013 hPa)
Melting Pt: 321 °C
Density: 8.642 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,II
Merck Index: 12,01649

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR