Niobium(V) oxide ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diniobium pentaoxide, Niobium pentoxide
11365-22
11365-36
11365-A3
CAAA11365-22EA
176.96
CAD
CAAA11365-22
CAAA11365-36
CAAA11365-A3
Niobium(V) oxide ≥99.5% (metals basis)
Niobium (V) oxide
As an intermediate and in electronics
Purity: 99.5%
CAS No.: 1313-96-8
Molecular Formula: Nb2O5
Formula Weight: 265.81
Physical Form: -100 Mesh Powder
Density: 4.6
Melting Point: 1520°
Solubility: Insoluble in water. Soluble in HF and alkalies
Merck Reference: 14,6559
MDL No.: MFCD00011128
As an intermediate and in electronics
Formula:
Nb₂O₅ Melting Pt: 1520 °C Density: 4.6 Storage Temperature: Ambient |
MDL Number:
MFCD00011128 CAS Number: 1313-96-8 EINECS: 215-213-6 Merck Index: 14,6559 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...