Silver ≥99.9985% (metals basis), rods, Premion®, Ø 12.5 mm (0.492 in)

Supplier: Thermo Scientific Chemicals

11356-BP 11356-BS
CAAA11356-BPEA 358.66 CAD
CAAA11356-BP CAAA11356-BS
Silver ≥99.9985% (metals basis), rods, Premion®, Ø 12.5 mm (0.492 in)
Silver

MDL: MFCD00003397


Purity: 99.9985% CAS No.: 7440-22-4 Physical Form: ≈12.87g/cm Odor: Characteristic MDL No.: MFCD00003397

Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR