Cadmium ≥99.99% (metals basis), powder -200 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
11353-06
11353-14
011353-22
CAAA11353-06EA
123.71
CAD
CAAA11353-06
CAAA11353-14
AA1135322
Cadmium ≥99.99% (metals basis), powder -200 mesh
Cadmium
MDL: MFCD00010914
Purity: 99.99%
CAS No.: 7440-43-9
Appearance: Silvery white
UN No.: 3179
MDL No.: MFCD00010914
Formula:
Cd Boiling Pt: 767 °C (1013 hPa) Melting Pt: 321 °C Density: 8.64 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010914 CAS Number: 7440-43-9 EINECS: 231-152-8 UN: 3179 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...