Cadmium ≥99.9997% (metals basis), shot, Puratronic® 1-3 mm (0.04-0.1 in)

Supplier: Thermo Scientific Chemicals
Danger

11351-18 11351-30 11351-A1
CAAA11351-18EA 363.4 CAD
CAAA11351-18 CAAA11351-30 CAAA11351-A1
Cadmium ≥99.9997% (metals basis), shot, Puratronic® 1-3 mm (0.04-0.1 in)
Cadmium

MDL: MFCD00010914


Purity: 99.9997% CAS No.: 7440-43-9 MDL No.: MFCD00010914

Formula: Cd
Boiling Pt: 765 °C (1013 hPa)
Melting Pt: 321 °C
Density: 8.642 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,II
Merck Index: 12,01649

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR