Antimony ≥99.999% (metals basis), shot ≤ 6 mm (0.2 in)

Supplier: Thermo Scientific Chemicals
Warning

11348-14 11348-22 11348-36
CAAA11348-14EA 170.64 CAD
CAAA11348-14 CAAA11348-22 CAAA11348-36
Antimony ≥99.999% (metals basis), shot ≤ 6 mm (0.2 in)
Antimony

MDL: MFCD00134030


Purity: 99.999% CAS No.: 7440-36-0 Odor: Odorless MDL No.: MFCD00134030

Formula: Sb
Boiling Pt: 1637 °C (1013 hPa)
Melting Pt: 630 °C
Density: 6.684 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1,III
Merck Index: 12,00733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR