Cobalt(II) chloride hexahydrate ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
11344-30
11344-A1
CAAA11344-30EA
154.05
CAD
CAAA11344-30
CAAA11344-A1
Cobalt(II) chloride hexahydrate ≥99.9% (metals basis)
Cobalt (II) chloride hexahydrate
MDL: MFCD00149652
Notes: m.p. 52-56°C -4H2O, 100°C -5H2O, 120-140°C -6H2O
Fieser: 1,155 6,127 10,101 14,99 15,97 18,107 19,104 20,115 21,142
Purity: 99.9%
CAS No.: 7791-13-1
Molecular Formula: Cl2Co.6H2O
Formula Weight: 237.93 (129.84anhy)
Physical Form: Crystalline
Density: 1.924
Appearance: Red-purple
Odor: Odorless
pH: 4.9
Melting Point: 87°
Merck Reference: 14,2437
UN No.: 3077
MDL No.: MFCD00149652
Formula:
CoCl₂·6H₂O Boiling Pt: 110 °C (1013 hPa) Melting Pt: 87 °C Density: 1.924 Storage Temperature: Ambient |
MDL Number:
MFCD00149652 CAS Number: 7791-13-1 EINECS: 231-589-4 UN: 3077 ADR: 9,III Merck Index: 14,2437 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...