Cobalt(II) carbonate (≥45% Co) ≥99% (metals basis) Co ≥ 45%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11343-22
11343-36
CAAA11343-22EA
135.56
CAD
CAAA11343-22
CAAA11343-36
Cobalt(II) carbonate (≥45% Co) ≥99% (metals basis) Co ≥ 45%
Cobalt (II) carbonate
In ceramics, manufacturing Co compounds, pigments, salts
Purity: 99%
CAS No.: 513-79-1
Molecular Formula: CoCO3
Formula Weight: 118.94
Physical Form: Powder
Density: 4.13
Melting Point: dec.
Solubility: Insoluble in water, alcohol, methyl acetate
Merck Reference: 14,2436
UN No.: 3077
MDL No.: MFCD00010940
In ceramics, manufacturing Co compounds, pigments, salts
Formula:
CCoO₃ Melting Pt: Dec. Density: 4.13 Storage Temperature: Ambient |
MDL Number:
MFCD00010940 CAS Number: 513-79-1 EINECS: 208-169-4 UN: 3077 ADR: 9,III Merck Index: 14,2436 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...