Cobalt(II) nitrate hexahydrate ≥97.7%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cobalt dinitrate hexahydrate, Cobaltous nitrate hexahydrate

11341-14 11341-30 11341-A1 11341-A7
CAAA11341-14EA 72.36 CAD
CAAA11341-14 CAAA11341-30 CAAA11341-A1 CAAA11341-A7
Cobalt(II) nitrate hexahydrate ≥97.7%
Cobalt (II) nitrate hexahydrate

MDL: MFCD00149647 Notes: greater than 74°C dec. to oxide


Purity: 97.7% min CAS No.: 10026-22-9 Molecular Formula: Co(NO3)2.6H2O Formula Weight: 291.04 (182.94anhy) Physical Form: Crystalline Density: 1.87 Refractive Index: 1.52 Appearance: Red Odor: Odorless Melting Point: 55-56° Merck Reference: 14,2444 UN No.: 1477 MDL No.: MFCD00149647

Formula: Co(NO₃)₂·6H₂O
Melting Pt: 55…56 °C
Density: 1.87
Storage Temperature: Ambient
MDL Number: MFCD00149647
CAS Number: 10026-22-9
EINECS: 233-402-1
UN: 1477
ADR: 5.1,III
Merck Index: 14,2444

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR