Cobalt(II) sulfate hydrate Co ≥ 20,8%, REacton®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cobalt sulfate hydrate, Cobaltous sulfate hydrate, Cobaltous sulphate hydrate, Cobalt sulphate hydrate

11340-30 11340-A1
CAAA11340-30EA 148.05 CAD
CAAA11340-30 CAAA11340-A1
Cobalt(II) sulfate hydrate Co ≥ 20,8%, REacton®
Cobalt (II) sulphate hydrate

Ceramics, pigments, glazes, plating baths, catalyst, storage batteries, paint/ink drier


CAS No.: 60459-08-7 Molecular Formula: CoO4S.xH2O Formula Weight: 154.99(anhy) Physical Form: Crystalline Appearance: Red-orange Solubility: Soluble in water. Slightly soluble in methanol and ethanol MDL No.: MFCD00149657


Ceramics, pigments, glazes, plating baths, catalyst, storage batteries, paint/ink drier

Formula: CoO₄S·H₂O
MW: 173.01
Storage Temperature: Ambient
MDL Number: MFCD00149657
CAS Number: 60459-08-7
EINECS: 233-334-2
UN: 3077
ADR: 9 III
Merck Index: 12,02510

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR