Tantalum(V) oxide ≥99.85% (metals basis)
Supplier: Thermo Scientific Chemicals
11336-14
11336-22
11336-36
CAAA11336-14EA
214.88
CAD
CAAA11336-14
CAAA11336-22
CAAA11336-36
Tantalum(V) oxide ≥99.85% (metals basis)
Tantalum (V) oxide
MDL: MFCD00011254
Purity: 99.85%
CAS No.: 1314-61-0
Molecular Formula: Ta2O5
Formula Weight: 441.89
Physical Form: -325 Mesh Powder
Density: 8.2
Melting Point: 1800°
Merck Reference: 14,9056
MDL No.: MFCD00011254
Formula:
O₅Ta₂ Melting Pt: 1800 °C Density: 8.2 Storage Temperature: Ambient |
MDL Number:
MFCD00011254 CAS Number: 1314-61-0 EINECS: 215-238-2 Merck Index: 14,9056 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...