Cerium trinitrate hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cerous chloride hexahydrate, Cerium (III) nitrate hexahydrate

11330-06 11330-18 11330-30
CAAA11330-06EA 69.99 CAD
CAAA11330-06 CAAA11330-18 CAAA11330-30
Cerium trinitrate hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Cerium trinitrate hexahydrate

MDL: MFCD00149631


Purity: 99.99% CAS No.: 10294-41-4 Molecular Formula: CeN3O9.6H2O Formula Weight: 434.23 (326.13anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic pH: 3 Merck Reference: 14,2000 UN No.: 1477 MDL No.: MFCD00149631

Formula: Ce(NO₃)₃·6H₂O
MW: 434.22 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 39 °C
Density: 2.377 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149631
CAS Number: 10294-41-4
EINECS: 233-297-2
UN: 1477
ADR: 5.1,III
Merck Index: 14,2000

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR