Cerium(IV) oxide ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ceric oxide, Cerium dioxide

11327-14 11327-22 11327-36
CAAA11327-14EA 114.55 CAD
CAAA11327-14 CAAA11327-22 CAAA11327-36
Cerium(IV) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Cerium (IV) oxide

MDL: MFCD00010933


Purity: 99.99% CAS No.: 1306-38-3 Molecular Formula: CeO2 Formula Weight: 172.12 Physical Form: 14 Micron Powder Density: 7.132 Appearance: Pale yellow Odor: Odorless Melting Point: 2600° Merck Reference: 14,1989 MDL No.: MFCD00010933

Formula: CeO₂
MW: 172.11 g/mol
Boiling Pt: 3468 °C (1013 hPa)
Melting Pt: 2600 °C
Density: 7.132
Storage Temperature: Ambient
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4
Merck Index: 14,1989

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR