Cerium triacetate sesquihydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cerium (III) acetate sesquihydrate, Acetic acid cerium (III) salt sesquihydrate

11324-09 11324-18 11324-30
CAAA11324-09EA 80.42 CAD
CAAA11324-09 CAAA11324-18 CAAA11324-30
Cerium triacetate sesquihydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Cerium triacetate sesquihydrate

Cerium(III) acetate is used in combination with bromide ion, catalyzes the liquid-phase auto-oxidation of cresols. It is used in glass manufacturing, glass Polishing, optical Glass making and polishing, pigments, pharma, Speciality chemicals electronic, oaint and driers, dyes and pigments, paper industries etc.


Purity: 99.99% CAS No.: 537-00-8 Molecular Formula: Ce(C2H3O2)3.1.5H2O Formula Weight: 344.28 (317.26anhy) Physical Form: Crystalline Aggregates Melting Point: 308° dec. MDL No.: MFCD00051060

Formula: Ce(C₂H₃O₂)₃·1.5H₂O
MW: 344.28 g/mol (317.26 anhy)
Melting Pt: 308 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051060
CAS Number: 537-00-8
EINECS: 208-654-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR